Methanol ≥99.8% ACS, VWR Chemicals BDH®
Catalog #: BDH1135-204L
Supplier: VWR International
CAS Number: 67-56-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Steel drum
- Size:204 L
- Supplier No.:PL230ZA-204
Specifications
Specification Test Results
- Appearance:Clear
- Water (K. F.):Less than 0.10%
- Residue:Less than 0.001% (0.0010 g/125 mL)
- GC Purity:Greater than 99.8%
- Solubility in water:Passes test (Solution should be as clear as an equal volume of water)
- Color (APHA):Less than 10
- Titrable acid:Less than 0.3 meq/g (No more than 0.50 mL of 0.01N sodium hydroxide)
- Titrable base:Less than 0.2 meq/g (No more than 0.40 mL of 0.01N hydrochloric acid)
- Carbonyl compounds (as acetone, formaldehyde, and acetaldehyde):Less than 0.001% each
- Substances darkened by sulfuric acid:Passes test (Solution should be colorless, or have no more color than the acid or methanol before mixing)
- Substances reducing permanganate:Passes test (Pink color should not be entirely discharged)
- VWR Chemicals BDH®, Methanol ≥99.8% ACS:
Specification Test Results
About this item
Synonyms: Hydroxy methane,Carbinol,Wood alcohol
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,8
Flash Pt:11 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 64.6 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –98 °C
:Flash Pt:
:Flash Pt: 11 °C (closed cup)
:M.W. 32.04 g/mol
Merck Index:13,05984
EINECS:200-659-6
:64.6
Storage Temperature:Ambient
UN:1230
REACH:01-2119433307-44
:3
:II
:11 °C
:GHS02
:GHS06
:GHS08
Customers Who Bought This Also Bought