Order Entry
ContactUsLinkComponent
MEDIUM LB BROTH MILLER TC GRADE 500G
VWR® Premixed LB Broth
Catalog #: 97064-114
Supplier:  VWR
CAS Number:  
MEDIUM LB BROTH MILLER TC GRADE 500G
VWR® Premixed LB Broth
Catalog #: 97064-114
Supplier:  VWR
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Miller Formulation
  • Size:
    500 g
  • Cat. no.:
    97064-114
  • Supplier No.:
    J106-500G

Specifications

About this item

Lysogeny broth (LB) is a nutritionally rich medium used for the growth of bacteria.

Lysogeny broth (LB) is a nutritionally rich medium used for the growth of bacteria. LB continues to be one of the most common media used for maintaining and cultivating recombinant strains of Escherichia coli. Powdered LB is available in premixed Miller or Lennox formulations and in Ready-Pack™ pouches which prepare 1L of media.

  • Save Time Using Premixed and Ready-to-Use Media
  • Homogenous and High Quality Powders

Lysogeny broth (LB) is a nutritionally rich medium used for the growth of bacteria. LB continues to be one of the most common media used for maintaining and cultivating recombinant strains of Escherichia coli. Powdered LB is available in premixed Miller or Lennox formulations and in Ready-Pack™ pouches which prepare 1L of media.

Customers Who Bought This Also Bought