Specifications
- Description:NUCLEASELIMINATOR™
- Size:100 mL
- Supplier No.:E891-100ML
Specifications
About this item
DNase and RNase contamination ruins expensive samples and time-consuming protocols in research laboratories every day. NucleasEliminator™ can help protect your investment by removing and deactivating even high concentrations of nuclease contamination from glass or plastic surfaces.
- Effectively removes and deactivates nuclease contamination even at high concentrations
- Safe on all laboratory glass and plastic ware
- Stable at room temperature for at least two years
Simply wipe, immerse or fill the surface or vessel to be decontaminated with NucleasEliminator™, incubate 5 minutes at room temperature and rinse thoroughly with nuclease-free water. NucleasEliminator™ is safe to use on all surfaces at the laboratory bench ranging from glassware, microcentrifuge tubes, pipette tips and electrophoresis units. NucleasEliminator™ is supplied as a ready-to-use liquid. A 100 ml bottle will treat up to 100 microcentrifuge tubes or 25 agarose electrophoresis casting trays.