VWR® EDTA, Biotechnology Grade
Catalog #: 97062-656
Supplier: VWR
CAS Number:
Specifications
- Size:500 mL
- Supplier No.:E177-500ML
Specifications
About this item
Customers Who Bought This Also Bought
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
66,85 $ to 318,03 $
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
635,20 $ to 2 541,50 $