Tricine, Ultrapure
Catalog #: 97062-642
Supplier: VWR
CAS Number: 5704-04-1
Specifications
- Packaging:HDPE bottle
- Size:100 g
- Cat. no.:97062-642
- Supplier No.:E170-100G
Specifications
Specification Test Results
- DNase:none detected
- Lead:= 0.001 %
- Melting Point:182°C - 188°C
- pKa @25°C:8.1 - 8.2
- Protease:none detected
- Purity:= 99 %
- RNase:none detected
- Solubility (10%, Water):Pass
- Tricine, Ultrapure:
Specification Test Results
About this item
Synonyms: 2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid,Tricine buffer
Melting Pt:187 °C
Density:1.523 g/cm³ (19.85 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 187 °C
:Flash Pt:
:Flash Pt:
:M.W. 179.17 g/mol
Merck Index:13,09724
Storage Temperature:Ambient
EINECS:227-193-6
Customers Who Bought This Also Bought