Bovine Serum Albumin (BSA)
Catalog #: 97062-508
Supplier: VWR
CAS Number:
Specifications
- Packaging:HDPE bottle
- Source:Serum
- Species:Bovine
- CAS No.:9048-46-8
90604-29-8 - Size:5 g
- Storage conditions:Refrigerator
- Protein synonyms:BSA|Fraction V|Cohn Fraction V
- Protein/peptide name:Bovine Serum Albumin
- Purity:99.0%
- Colour:light yellow
- pH:7% Water, 5.0-5.4 at 25°C
- Grade:for biotechnology
- Supplier No.:0903-5G
Specifications
About this item
Crystalline. Prepared by a cold alcohol isolation procedure. This salt-free preparation is the purest material ideal for sensitive biochemical and diagnostic assays. DNase- and RNase-free, virtually free of globulins and other interfering contaminants.
Customers Who Bought This Also Bought