To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Bottle, amber
- CAS No.:9003-99-0
- Size:25000 units
- Storage conditions:Freezer
- MDL No.:MFCD00071339
- Enzyme name:Peroxidase
- Tested applications:for biochemistry
- Supplier No.:0343-25,000U
Specifications
About this item
HRP catalyzes the decomposition of hydrogen peroxide in solution causing the oxidation of a number of substrates.
Used extensively as a label for immunohistological assays and ELISA. Its exceptional stability in powdered and solution form enhances its desirability as an enzyme conjugate.