EDTA (ethylenediamine tetraacetic acid) free acid, Ultrapure
Catalog #: 97061-406
Supplier: VWR
CAS Number: 60-00-4
Specifications
- Packaging:HDPE bottle
- Size:500 g
- Cat. no.:97061-406
- Supplier No.:0322-500G
Specifications
Specification Test Results
- DNase:none detected
- Heavy Metals (as Pb):= 0.001%
- Insolubles (5%, 1N NaOH):Pass
- Iron:= 0.005%
- Magnesium:= 0.0005%
- Nitrilotriacetic Acid:= 0.1%
- pH (5%, Water) @25°C:2.5 - 3.5
- Protease:none detected
- Purity:= 99.5%
- Residue after Ignition:= 0.2%
- RNase:none detected
- EDTA (ethylenediamine tetraacetic acid) free acid, Ultrapure:
Specification Test Results
About this item
Synonyms: Kestranal® A,OmniPur® EDTA,Edetic acid,Ethylenediaminetetraacetic acid,Ethylenedinitrilotetra-acetic acid,2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid,(Ethylenedinitrilo)tetraacetic acid
Boiling Pt:614 °C (1013 hPa)
Melting Pt:240 °C
Density:0.86 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
Flash Pt:>100 °C
:Boiling Pt:
:Boiling Pt: 614 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 240 °C
:Flash Pt:
:Flash Pt: >100 °C
:M.W. 292.25 g/mol
Merck Index:13,03546
:EDTA (ethylenediamine tetraacetic acid)
EINECS:200-449-4
REACH:01-2119486399-18
:>100 °C
:GHS07
Storage Temperature:Ambient
Customers Who Bought This Also Bought