SP Wilmad-LabGlass Thin Walled High Throughput NMR Tubes, SP Industries
Catalog #: 89041-318
Supplier: SP Industries
Specifications
- Length:203 mm (8")
- O.D.:4.94665±0.01905 mm
- Wall thickness:0.43 mm (0.0169")
- Frequency:High-Throughput
- Supplier No.:WG-1000-8
Specifications
About this item
Wilmad Thin Walled High Throughput NMR Tubes have an average camber of 60 microns to guarantee the clarity of the spectrum for small molecule (MW<250) samples up to 600 MHz.
- Camber and concentricity values listed are the Total Indicator Reading (TIR)
- Designed for routine use in most low to mid field NMR spectrometers
- One of the best OD tolerances in the industry
- Guaranteed to fit tightly in spinner turbine
- Made from ASTM E438 Type 1 Class B glass
- 100% inspected for surface defects and physical dimension to ensure the success of your experiments
Note: For NMR experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, please refer to Wilmad Precision NMR Tubes.
Delivery: WG-1000-4-SJ and WG-1000-7-SJ are supplied with authentic Bruker SampleJet caps, all others come with a disposable NMR cap.
Customers Who Bought This Also Bought