Order Entry
ContactUsLinkComponent
Graduated Centrifuge Tube
Quick-Prep™ Urinalysis Kits, Globe Scientific
Catalog #: 60819-982
Supplier:  Globe Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Graduated Centrifuge Tube
Quick-Prep™ Urinalysis Kits, Globe Scientific
Catalog #: 60819-982
Supplier:  Globe Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Capacity:
    12 ml
  • Ø:
    20 mm
  • Description:
    Flared Top Urine Tube, Ames Type, Graduated, Round Bottom
  • Height:
    115 mm
  • Material:
    Polystyrene
  • Supplier No.:
    112017

Specifications

About this item

Quick-Prep™ is a tube and pipette system that isolates 1mL of urine sediment for microscopic examination

Quick-Prep™ kits have been designed for use with the Quick-Read™ (10-place) urinalysis slide.

The kit includes specially designed transfer pipette and 12mL urine centrifuge tube. This system is used to isolate 1mL of urine sediment stain and allow easy pour-off of the balance.

Customers Who Bought This Also Bought