D-(+)-Glucose for biotechnology
Catalog #: 97061-164
Supplier: VWR
CAS Number: 50-99-7
Specifications
- Size:1 kg
- Supplier No.:0188-1KG
Specifications
Specification Test Results
- Arsenic:= 0.00004%
- Chloride:= 0.01%
- DNase:none detected
- Heavy Metals (as Pb):= 0.0005%
- Insolubles:= 0.005%
- Iron:= 0.0005%
- Loss on Drying:= 0.2%
- Protease:none detected
- Purity:= 99.5%
- Residue on Ignition:= 0.02%
- RNase:none detected
- Specific Rotation:52.5°- 54 °
- Sulfate & Sulfite:= 0.005%
- D-(+)-Glucose for biotechnology:
Specification Test Results
About this item
Synonyms: Corn sugar,Glucose,D-(+)-Dextrose,(2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal,Glucolin,Dextrose
Melting Pt:146 °C
Density:1.54 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 146 °C
:Flash Pt:
:Flash Pt:
:M.W. 180.16 g/mol
Merck Index:14,04459
:D-(+)-Glucose
Storage Temperature:Ambient
EINECS:200-075-1
Customers Who Bought This Also Bought