KLEENGUARD® A10 Light Duty Lab Coat, Kimberly-Clark Professional®
Catalog #: 10837-436
Supplier: Kimberly-Clark
Specifications
- Clothing Size:L
- Garment application:Laboratory
- Color:White
- Description:Snap front, elastic wrists, no pocket
- Environmentally Preferable:Waste Reducing
- Cat. no.:10837-436
- Supplier No.:40103
Specifications
About this item
Lightweight, breathable polypropylene fabric keeps workers cool and comfortable
- Lightweight, breathable polypropylene fabric
- Combination of price and protection for nonhazardous environments
These lab coats provide the right combination of price and protection for non-hazardous environments.
Customers Who Bought This Also Bought