To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:2000 µg/ml each
- Matrix:Dichloromethane
- Description:SV Internal Standard Mix
- Size:1 Ampule
- Cat. no.:RK31206
- Supplier No.:31206
Specifications
About this item
Standards are packaged in methylene chloride in a 1 mL ampoule.
- High purity for consistent results
- Free quality assurance data package available online
- Highest concentrations commercially available.
Mixture contains Acenaphthene-d10, Chrysene-d12, 1,4-Dichlorobenzene-d4, Naphthalene-d8, Perylene-d12, and Phenanthrene
US Environmental Protection Agency (EPA) Method 8270D outlines the analysis of semivolatile organic pollutants in solid waste, soil, water, and air matrices, using gas chromatography/mass spectrometry (GC/MS). Update IVA of the third edition of SW-846—Test Methods for Evaluating Solid Waste, Physical/Chemical Methods—includes EPA Method 8270D, in which there were no major revisions from EPA Method 8270C.