ContactUsLinkComponent
Low-Pressure, Slip-on Inlet Filters
Low-Pressure Slip-On Inlet Filter for Mobile Phase Reservoir, Restek
Catalog #: RK25008
Supplier:  Restek
Low-Pressure, Slip-on Inlet Filters
Low-Pressure Slip-On Inlet Filter for Mobile Phase Reservoir, Restek
Catalog #: RK25008
Supplier:  Restek
Specifications
  • Description:
    Low-Pressure Inlet Filter, 1/8" 10 µm Slip-On
  • Cat. no.:
    RK25008
  • Supplier No.:
    25008
Specifications
About this item
A 316 stainless steel tip with a Tefzel® collar seals to a corrosion-resistant 316 stainless steel filter element.
The slip-on filter easily attaches to the pump inlet line, without the use of wrenches. The universal ¹/₈" Ext.Ø tip accommodates standard PTFE tubing inner diameters. The cylindrical filter is standard 10 µm porosity. Fits Altex, ISCO, LDC, Varian, Waters, PerkinElmer, and other pumps.
Customers Who Bought This Also Bought