Order Entry
ContactUsLinkComponent
GRAPHITE SEALING RING PK2
Graphite Sealing Ring for Thermo Scientific TRACE, 8000, 8000 TOP & Focus SSL instruments; fits 8 mm OD liners, Restek
Catalog #: RK21899
Supplier:  Restek
GRAPHITE SEALING RING PK2
Graphite Sealing Ring for Thermo Scientific TRACE, 8000, 8000 TOP & Focus SSL instruments; fits 8 mm OD liners, Restek
Catalog #: RK21899
Supplier:  Restek

Specifications

  • Description:
    Graphite Sealing Ring
  • Cat. no.:
    RK21899
  • For Use With:
    for Thermo Scientific TRACE, 8000, 8000 TOP & Focus SSL
  • Supplier No.:
    21899

Specifications

About this item

Graphite sealing rings are ideal for use with Thermo Scientific TRACE, 8000, 8000 TOP, and Focus SSL instruments.

  • Compatible with 8 mm OD liners
  • Durable graphite
  • Max operating temperature of 450°C
Customers Who Bought This Also Bought