Aniline ≥99.0% (by GC, corrected for water content), AR®, Macron Fine Chemicals™
Catalog #: MK358404
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 62-53-3
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber, narrow mouth
- Size:500 mL
- Cat. no.:MK358404
- Supplier No.:3584-04
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Assay (C₆H₅NH₂) (by GC, corrected for water):≥99.0 %
- Color (APHA) (at time of packaging):≤250
- Residue after Ignition:≤0.005 %
- Chlorobenzene (C₆H₅Cl):≤0.01 %
- Hydrocarbons:Passes Test
- Nitrobenzene (C₆H₅NO₂):Passes Test
- NOTE: This reagent darkens to a reddish-brown color on storage.:
Specification Test Results
About this item
Synonyms: Aminobenzene
Boiling Pt:184 °C (1013 hPa)
Melting Pt:–6.2 °C
Density:1.019 g/cm³ (20 °C)
ADR:6.1,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
:Management of Change (MOC) category = N
Flash Pt:76 °C
:Boiling Pt:
:Boiling Pt: 184 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –6.2 °C
:Flash Pt:
:Flash Pt: 76 °C
:M.W. N/A N/A
Merck Index:13,00661
:Aniline
Storage Temperature:Ambient
EINECS:200-539-3
UN:1547
:6.1
:II
:GHS06
:GHS08
:GHS09
:M.O.C. Category N
Customers Who Bought This Also Bought