Acetic acid glacial ≥99.7% (by GC, corrected for water content), BAKER ANALYZED® ACS aldehyde-free, J.T.Baker®
Catalog #: JT9508-2
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 64-19-7
MOC: Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, clear in a single shipper
- Size:500 ml
- Supplier No.:9508-02
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- IMPORTANT: Material will freeze if stored below 17 °C (63°F).:
- Assay (by GC, corrected for water):≥99.7 %
- Color (APHA):≤10
- Dilution Test:Passes Test
- Residue after Evaporation:≤0.001 %
- Acetic Anhydride ((CH₃CO)₂O):≤0.01 %
- Chloride (Cl):≤1 ppm
- Sulfate (SO₄):≤1 ppm
- Trace Impurities - Iron (Fe):≤0.2 ppm
- Substances Reducing Dichromate:Passes Test
- Substances Reducing Permanganate:Passes Test
- Titrable Base (meq/g):≤0.0004
- Acetaldehyde:≤0.005 %
- Sensitivity:Passes Test
- Trace Impurities - Arsenic and Antimony (as As):≤50.0 ppb
- Trace Impurities - Boron (B):≤100.0 ppb
- Trace Impurities - Chromium (Cr):≤200.0 ppb
- Trace Impurities - Cobalt (Co):≤100.0 ppb
- Trace Impurities - Copper (Cu):≤100.0 ppb
- Trace Impurities - Heavy Metals:≤0.5 ppm
- Trace Impurities - Lead (Pb):≤300.0 ppb
- Trace Impurities - Manganese (Mn):≤200.0 ppb
- Trace Impurities - Nickel (Ni):≤100.0 ppb
- Trace Impurities - Potassium (K):≤300.0 ppb
- Trace Impurities - Tin (Sn):≤100 ppb
- Trace Impurities - Titanium (Ti):≤300.0 ppb
- Trace Impurities - Zinc (Zn):≤200.0 ppb
- Meets Reagent Specifications for testing USP/NF monographs:
- J.T.Baker®, Acetic acid glacial ≥99.7% (by GC, corrected for water content), BAKER ANALYZED® ACS aldehyde-free:
Specification Test Results
About this item
Synonyms: Vinegar acid,Methanecarboxylic acid
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1.05 g/cm³ (20 °C)
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,7
:Management of Change (MOC) category = N
Flash Pt:38.5 °C
:Boiling Pt:
:Boiling Pt: 118 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 17 °C
:Flash Pt:
:Flash Pt: 38.5 °C
:M.W. 60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
REACH:01-2119475328-30
:8
:II
:GHS02
:GHS05
:GHS07
:M.O.C. Category N
Customers Who Bought This Also Bought