Talc, powder, Multi-Compendial, J.T.Baker®
Catalog #: JT4100-1
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 14807-96-6
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Poly bottle, white, blue T/E cap
- Size:500 g
- Cat. no.:JT4100-1
- Supplier No.:4100-01
Specifications
Specification Test Results
- GMP Manufactured Product:
- Meets U.S.P Requirements:
- Meets B.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Identification C:Passes Test
- Microbial Limit - Total Aerobic Bacterial Count:≤100 cfu/g
- Microbial Limit - Total Molds and Yeasts:≤50 cfu/g
- USP - Acidity and Alkalinity:Passes Test
- USP - Loss on Ignition:≤7.0 %
- USP - Water-Soluble Substances:≤0.1 %
- USP - Iron (Fe):≤0.25 %
- USP - Lead (Pb):≤0.001 %
- USP - Calcium (Ca):≤0.9 %
- USP - Aluminum (Al):≤2.0 %
- USP - Absence of Asbestos (Method A):Passes Test
- USP - Magnesium (as MgO):17.0 - 19.5 %
- EP/BP - Free from Asbestos:Passes Test
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Water-Soluble Substances:≤0.2 %
- EP/BP - Aluminum (Al):≤2.0 %
- EP/BP - Calcium (Ca):≤0.90 %
- EP/BP - Iron (Fe):≤0.25 %
- EP/BP - Lead (Pb):≤10.0 ppm
- EP/BP - Magnesium (Mg):17.0 - 19.5 %
- EP/BP - Loss on Ignition:≤7.0 %
- EP/BP - Total Aerobic Microbial Count, cfu/g:≤1000
- EP/BP - Total Yeast and Mold Count, cfu/g:≤100
- No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
- Suitable for Oral Administration:
Specification Test Results
About this item
Synonyms: Magnesium silicate monohydrate,Soapstone
Melting Pt:1200 °C
Density:2.58…2.83 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: Min. 1200 °C|Melting Pt: 1200 °C
:Flash Pt:
:Flash Pt:
:M.W. 379.27 g/mol
Merck Index:13,09127
EINECS:238-877-9
:GHS08
Storage Temperature:Ambient
:M.O.C. Category R
Customers Who Bought This Also Bought