1-Hexanesulfonic acid sodium salt ≥98.0% for HPLC, for ion pair chromatography, J.T.Baker®
Catalog #: JT2175-5
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 2832-45-3
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber, wide mouth, black T/E cap
- Size:25 g
- Supplier No.:2175-05
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Assay (as C₆H₁₃SO₃Na) (by acidimetry):≥98.0 %
- UV Absorbance of 0.25 M Solution - 250 nm:≤0.050
- UV Absorbance of 0.25 M Solution - 240 nm:≤0.050
- UV Absorbance of 0.25 M Solution - 230 nm:≤0.050
- UV Absorbance of 0.25 M Solution - 220 nm:≤0.060
- UV Absorbance of 0.25 M Solution - 210 nm:≤0.080
- UV Absorbance of 0.25 M Solution - 200 nm:≤0.200
- Appearance (clear, colorless, mobile liquid):
- J.T.Baker®, 1-Hexanesulfonic acid sodium salt ≥98.0% for HPLC, for ion pair chromatography:
Specification Test Results
About this item
Synonyms: Sodium hexane-1-sulfonate,Sodium hexane-1-sulphonate,Sodium hexanesulfonate,Sodium hexanesulphonate,Sodium 1-hexanesulfonate
Formula:C₆H₁₃O₃SNa
Melting Pt:300 °C
MDL Number:MFCD00007542
CAS Number:2832-45-3
:Storage Temperature: Ambient
:Length
:Width
:Height
:Diameter
:Thickness
:98,0
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 300 °C
:Flash Pt:
:Flash Pt:
:M.W. 188.22 g/mol
Storage Temperature:Ambient
EINECS:220-601-3
:M.O.C. Category N
Customers Who Bought This Also Bought