γ-(N-Morpholino)propanesulfonic acid (MOPS), Ultrapure
Catalog #: 97062-160
Supplier: VWR
CAS Number: 1132-61-2
Specifications
- Size:500 g
- Cat. no.:97062-160
- Supplier No.:0670-500G
Specifications
Specification Test Results
- Abs.@260nm (0.1M, Water):= 0.05
- Abs.@280nm (0.1M, Water):= 0.05
- DNase:none detected
- RNase:none detected
- pKa @(25°C):7 - 7.4
- Protease:none
- Purity:99 %
- γ-(N-Morpholino)propanesulfonic acid (MOPS), Ultrapure:
Specification Test Results
About this item
Synonyms: 3-Morpholinopropane-1-sulfonic acid,3-(N-Morpholino)propanesulfonic acid,3-Morpholinopropane-1-sulphonic acid
Melting Pt:277 °C
:Length:
:Width:
:Height:
:Ø
:Thickness:
Flash Pt:110 °C (closed cup)
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 277 °C
:Flash Pt:
:Flash Pt: 110 °C (closed cup)
:M.W. 209.27 g/mol
Merck Index:12,06346
:γ-(N-Morpholino)propanesulphonic acid (MOPS)
EINECS:214-478-5
:110 °C
:GHS07
Storage Temperature:Ambient
Customers Who Bought This Also Bought