EDTA tetrasodium salt hydrate ≥99.5%, Ultra Pure Grade
Catalog #: 97061-294
Supplier: VWR
CAS Number: 13235-36-4
Specifications
- Packaging:HDPE bottle
- Size:500 g
- Supplier No.:0245-500G
Specifications
Specification Test Results
- Heavy Metals (as Pb):= 0.001 %
- Solubility (5%, Water):pass
- Iron:= 0.005 %
- Moisture (KF):12.4 % - 19.2 %
- Nitrilotriacetic Acid:= 0.1 %
- pH (1%, Water) @25°C:10.7 - 11.7
- Purity (Dry Basis):99.5 %
- EDTA tetrasodium salt hydrate ≥99.5%, Ultra Pure Grade:
Specification Test Results
About this item
Synonyms: Tetrasodium ethylenediaminetetraacetate hydrate,Kestranal® 4S hydrate,Ethylenedinitrilotetra-acetic acid tetrasodium salt hydrate,2,2',2'',2'''-(ethane-1,2-diylbis(azanetriyl))tetraacetate hydrate
Melting Pt:> 300 °C
Density:6.9 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: > 300 °C
:Flash Pt:
:Flash Pt:
:M.W. 380.17 g/mol (anhy)
Merck Index:12,03557
EINECS:200-573-9
:GHS06
:GHS07
Storage Temperature:Ambient
Customers Who Bought This Also Bought