Glycine for biotechnology
Catalog #: 97061-128
Supplier: VWR
CAS Number: 56-40-6
Specifications
- Size:1 kg
- Cat. no.:97061-128
- Supplier No.:0167-1KG
Specifications
Specification Test Results
- Abs.@260 nm (1M solution):≤0.1
- Abs.@280 nm (1M solution):≤0.1
- Chloride:= 0.01%
- DNase:none detected
- Heavy Metals (as Pb):= 0.002%
- Loss on Drying:= 0.2%
- pH (5%, Water) @25°C:5.9 - 6.4
- Protease:none detected
- Purity:= 99%
- RNase:none detected
- Solubility (10%, Water):Pass
- Sulfate:= 0.05%
- Thin Layer Chromatography (TLC):One Spot
- Glycine for biotechnology:
Specification Test Results
About this item
Synonyms: Aminoacetic acid,Glycocoll
Boiling Pt:181 °C (1013 hPa)
Melting Pt:240 °C
Density:1.64 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt: 181 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 240 °C
:Flash Pt:
:Flash Pt:
:M.W. 75.07 g/mol
Merck Index:13,04504
:Glycine
Storage Temperature:Ambient
EINECS:200-272-2
Customers Who Bought This Also Bought