Orthophosphoric acid ≥85.0-87.0%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog #: JT0260-2
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 7664-38-2
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, clear in a single shipper
- Size:500 ml
- Cat. no.:JT0260-2
- Supplier No.:0260-02
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- Assay (H₃PO₄) (by acidimetry):85.0 - 87.0 %
- Calcium (Ca):≤0.002 %
- Color (APHA):≤10
- Insoluble Matter:≤0.001 %
- ACS - Magnesium (Mg):≤0.002 %
- Sulfate (SO₄):≤12 ppm
- Volatile Acids (as CH₃COOH):≤0.001 %
- Reducing Substances:Passes Test
- Chloride (Cl):≤3 ppm
- Nitrate (NO₃):≤5 ppm
- Trace Impurities - Antimony (Sb):≤20.000 ppm
- Trace Impurities - Arsenic (As):≤0.500 ppm
- Trace Impurities - Iron (Fe):≤10 ppm
- Heavy Metals (as Pb):≤8 ppm
- Trace Impurities - Manganese (Mn):≤0.500 ppm
- Trace Impurities - Potassium (K):≤40.000 ppm
- Trace Impurities - Sodium (Na):≤200.000 ppm
- Exceeds A.C.S. Specifications:
- Meets Reagent Specifications for testing USP/NF monographs:
- J.T.Baker®, Orthophosphoric acid ≥85.0-87.0%, BAKER ANALYZED® ACS:
Specification Test Results
About this item
Synonyms: Phosphoric acid
Melting Pt:28 °C
Density:1.69 g/cm³
ADR:8,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:85,0-87,0
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 28 °C
:Flash Pt:
:Flash Pt:
:M.W. 98 g/mol
EINECS:231-633-2
:21
:158
UN:1805
:8
:III
:GHS05
:GHS07
Storage Temperature:Ambient
:M.O.C. Category N
:12,07500
:14,07344
Customers Who Bought This Also Bought