VWR® Ready Ladder™ 50 bp DNA Marker
Catalog #: 97063-302
Supplier: VWR
CAS Number:
Specifications
- Size:100 rxn
- Cat. no.:97063-302
- Application:Sufficient for 100 lanes
- Supplier No.:N746-100RXN
Specifications
About this item
10 fragments ranging from 50 to 500 bp for easy band identification
Ready-to-use formula pre-mixed with loading dye.
Customers Who Bought This Also Bought