Bovine Serum Albumin (BSA)
Catalog #: 97062-906
Supplier: VWR
CAS Number:
Specifications
- Packaging:HDPE Bottle
- Source:Serum
- Species:Bovine
- CAS No.:9048-46-8
90604-29-8 - Size:25 g
- Storage conditions:Refrigerator
- Protein synonyms:BSA|Fraction V|Cohn Fraction V
- Protein/peptide name:Bovine Serum Albumin
- Colour:light yellow
- Grade:for radioimmunoassay (RIA)
- Cat. no.:97062-906
- Supplier No.:E588-25G
Specifications
About this item
Customers Who Bought This Also Bought