To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- CAS No.:39450-01-6
- Size:100 mg
- Storage conditions:Freezer
- Enzyme name:Proteinase K
- Grade:Biotechnology Grade
- Supplier No.:0706-100MG
Specifications
About this item
Lyophilised powder
Proteinase K is useful for the inactivation of nucleases during the isolation of DNA and RNA. Active over a broad pH range. No DNase or RNase detected.
A non specific protease with a molecular weight of approximately 18 kDa. Exhibits high stability and activity in the presence of SDS, EDTA, and urea, as well as over a wide pH range. Activity >30 units/mg.