To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Ø:11 mm
- Length:40 mm
- Volume:1 ml
- Description:Serum Clot Activator
- Tube Dimensions:11 x 40 mm
- Cap Color:Red
- Supplier No.:450470
Specifications
About this item
Tubes are suitable for collection, transport, and processing of capillary blood samples from infants, toddlers, children, geriatric patients, and patients in intensive care.
Serum and plasma tubes may be used for testing parameters in clinical chemistry. Tubes with a gel separator contain an inert, acrylic barrier in the base of the tube with a specific gravity that lies between the clot/cells and serum/plasma. During centrifugation, the gel will shift to form a stable barrier between the clot/cells and the serum/plasma. The barrier remains stable for transportation. The EDTA tubes may be used for testing parameters in hematology.
- May be used to test clinical chemistry parameters
- Stable gel barrier for transport
- Available in a variety of formats for specific uses
Tubes are suitable for collection, transport, and processing of capillary blood samples from infants, toddlers, children, geriatric patients, and patients in intensive care.
Serum and plasma tubes may be used for testing parameters in clinical chemistry
Serum and plasma tubes may be used for testing parameters in clinical chemistry
The EDTA tubes may be used for testing parameters in hematology. Carrier tubes are designed to ensure compatibility with most common centrifuges and laboratory instruments. The blood collection capillary and funnel are used to simplify blood transfer.
Tubes with a gel separator contain an inert, acrylic barrier in the base of the tube with a specific gravity that lies between the clot/cells and serum/plasma. During centrifugation, the gel will shift to form a stable barrier between the clot/cells and the serum/plasma. The barrier remains stable for transportation. These application-specific tubes are available in a variety of fill volumes.