To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Ø:13 mm
- Length:100 mm
- Volume:6 ml
- Description:K2EDTA Tube
- Tube Dimensions:13 x 100 mm
- Ring Color:Black
- Cap Color:Lavender
- Supplier No.:456002
Specifications
About this item
Designed for use in the examination of whole blood in hematology applications, EDTA tubes feature interior walls that are coated with either K2EDTA or K3EDTA.
The EDTA tubes equipped with a gel separator contain a barrier gel in the base of the tube. During centrifugation, this gel forms a stable barrier between the serum and the blood cells, separating the serum from fibrin and cells.
- Gel separator forms stable barrier between serum and blood cells
- PET plastic construction is virtually unbreakable
- Color-coded rings in caps
Designed for use in the examination of whole blood in hematology applications, EDTA tubes feature interior walls that are coated with either K2EDTA or K3EDTA.
The EDTA binds calcium ions and blocks the coagualtion cascade
The EDTA binds calcium ions and blocks the coagualtion cascade
The EDTA tubes equipped with a gel separator contain a barrier gel in the base of the tube. During centrifugation, this gel forms a stable barrier between the serum and the blood cells, separating the serum from fibrin and cells.
VACUETTE® tubes are manufactured from virtually unbreakable PET plastic. These clear tubes are supplied with non-ridged pull caps that feature an exterior color-coded ring to indicate either the additive used or draw volume. A black ring indicates standard tubes, a yellow ring indicates the inclusion of a separation gel, and a white ring indicates a tube designed to accommodate pediatric draw.
Tubes are spray-dried with 1.2–2.0mg EDTA/1 mL of blood. Tubes can also be used in the closed mode with direct sampling analyzers.