Specifications
- Description:SB50 Grid Box
- Supplier No.:71135-01
Specifications
About this item
The clear cover can be rotated smoothly through 360 degrees once the slight initial resistance of the park position has been overcome exposing a maximum of 2 or 3 diamond shaped slots at any one time
The 50 diamond shaped slots have an alphanumeric referencing system. Each box is supplied with an index record card for additional information. The storage boxes are designed to be stacked, the base locating precisely over the face of another box. The material the bases are made from have been chosen due to their anti-static properties. The clear cover has self-lubricating properties.