Specifications
- Point Size:—
- Description:Diamond 60° Tip Retractable Scriber
- Cat. no.:100491-582
- Length:—
- Supplier No.:70036
- Diameter:—
Specifications
About this item
Diamond tip scribers are in wide use throughout the laboratory. It leaves a permanent marking on virtually any surface, including metal, glass, or plastic.
The retractable scriber is a high quality chrome pen that features appearance as well as utility. The diamond is mounted in a stainless steel tip and refills are available. The tip is finished at a 60° angle.
- Hexagonal aluminum pencil shaped body with a mill finish
- Natural diamond tip
- Protective cap included
- Straight and Angle tip scribers are available