Specifications
- Color:Blue
- Description:Plastic Forcep with Jaw-Grips
- Supplier No.:72971
Specifications
About this item
These plastic forceps are excellent for handling hot, cold, corrosive or contaminated objects. Made from blue polypropylene with interlocking teeth.
A catch locks tips tightly for use as a clamp. Scissor type handles with double action hinges makes this forcep a useful cryo tool, and great for handling stubs.