ContactUsLinkComponent
Lint-free cloth 9x9
Cloth; Lint Free, Twill Jean, Electron Microscopy Sciences
Catalog #: 100488-444
Minority or Woman-Owned Business Enterprise
Lint-free cloth 9x9
Cloth; Lint Free, Twill Jean, Electron Microscopy Sciences
Catalog #: 100488-444
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Color:
    White
  • Description:
    Lint-Free Cloth
  • L×W:
    9×9"
  • Supplier No.:
    71750
Specifications
About this item
A twill jean cotton wipe made from 100% cotton fibre that is woven in an extremely tight construction.
  • Highly absorbent
  • Thermal stability, good for high heat applications
  • Excellent wipe ability
  • Acid resistant and solvent compatible
  • Low metallic ion content
  • Low static generation
The extractable trace elements are effectively reduced in the cleaning process which removes impurities in the cotton as well as any starches or binders used in the manufacturing process.
Customers Who Bought This Also Bought