Order Entry
ContactUsLinkComponent
Laser Tough Tag
Chemically Inert Polyester Tags on Sheets, Electron Microscopy Sciences
Catalog #: 101413-822
Minority or Woman-Owned Business Enterprise
Laser Tough Tag
Chemically Inert Polyester Tags on Sheets, Electron Microscopy Sciences
Catalog #: 101413-822
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • For Use With:
    General purpose
  • Color:
    White
  • Description:
    Chemically inert polyester Tough-Tags on sheets
  • Material:
    Polyester
  • Packaging:
    64 labels/sheet
  • Dimensions:
    41 x 16 mm
  • Supplier No.:
    77528-W

Specifications

About this item

Tough-Tags® labels are mainly used for microcentrifuge tubes and other containers. These labels are made of chemically inert polyester sheets which resist most organic solvents and caustic agents.

  • Adhere to most plastics, glass and metals without cracking, peeling or degrading
  • Temperature resistance: −40 to 121 °C

Label sheets are specially top-coated to readily accept laser printing. Stay-flat sheets will not jam in laser printers. Tubes labeled with Tough-Tags® are easily slide in and out of centrifuge rotors without binding.

Customers Who Bought This Also Bought