To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Single Edge Carbon Steel Heavy Duty
- Thickness:0.012"
- Supplier No.:71964
Specifications
About this item
Single edge, carbon steel razor blades are available as regular type, PAL Extra Keen, GEM Extra Keen, and heavy duty.
- All blades are 11/2 x 3/4" in size
- Blades are 0.009" thick except heavy duty
- Heavy duty blades are 0.012" thick
Regular and PAL Extra Keen blades are available in a washed and degreased version, individually wrapped in a clamshell, therefore reducing the amount of foreign material on the blade. Clean room classified.