Ultragrade 19 Vacuum Pump Oil 4L
Catalog #: CA11010-294
Supplier: Edwards
Customers Who Bought This Also Bought
Hexane (mixture of isomers) ≥98.5% ACS for HPLC, Supelco®
Supplier: MilliporeSigma
$649.44 to $2,496.86
Masterflex® Luer Stopcock Fittings, Polycarbonate, Avantor®
Supplier: VWR International
$64.62 to $66.68
Tygon S3™ Laboratory Tubing, Formulation E-3603, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Life Sciences
$1,225.54
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$500.19 to $1,790.36
Disposable Base Mold, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$224.27 to $254.10