Orders LinkContactUsLinkComponent
Cheese Cloth, Electron Microscopy Sciences
Cheese Cloth, Electron Microscopy Sciences
Cheese Cloth, 60 yds.
Catalog # 100488-116
Supplier #:  71748-00
Cheese Cloth, Electron Microscopy Sciences
Catalog # 100488-116
Cheese Cloth, Electron Microscopy Sciences
Cheese Cloth, Electron Microscopy Sciences
Cheese Cloth, 60 yds.
Price
$262.85
Log in for availability
Each (180Feet) Roll of 180
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Cotton woven cloth is suitable for cleaning and polishing.
  • (44×36") 44 lines/inch horizontal
  • 36 lines/inch vertical
  • Supplied in 60 yard rolls 36" in width
This configuration allows to cut out the exact length desired.
Specifications
  • Description:
    Cheese Cloth
  • Length:
    60 yds.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)