Order Entry
ContactUsLinkComponent
CERAMIC PASTE ONLY 180MM
Liquid Nitrogen-Cooled Mortar, Electron Microscopy Sciences
Catalog #: 101413-004
Minority or Woman-Owned Business Enterprise
CERAMIC PASTE ONLY 180MM
Liquid Nitrogen-Cooled Mortar, Electron Microscopy Sciences
Catalog #: 101413-004
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Ceramic Pestle
  • Cat. no.:
    101413-004
  • Supplier No.:
    64472

Specifications

About this item

The Liquid Nitrogen-Cooled Mortar is for grinding frozen tissue samples without the hazard of liquid nitrogen droplets flying into personnel.

  • Insulated handle 4" (102 mm) is removable for saving space
  • Housing measures 61/2" (165 mm) in diameter, 41/2" high
  • Three suction cup-feet hold the housing

A stainless steel mortar is suspended by three brackets above a stainless steel bowl which holds the liquid nitrogen. The bowl is nested in a well insulated polyethylene housing.

Customers Who Bought This Also Bought