Order Entry
ContactUsLinkComponent
Super slip micro glasses 24x50-1oz
Superslip® Micro Glasses, Electron Microscopy Sciences
Catalog #: 100502-562
Minority or Woman-Owned Business Enterprise
Super slip micro glasses 24x50-1oz
Superslip® Micro Glasses, Electron Microscopy Sciences
Catalog #: 100502-562
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Numbering:
    #1
  • Volume:
    1 oz.
  • Description:
    Cover glasses, Super Slip, rectangular
  • L×W:
    50×24 mm
  • Packaging:
    80 pieces/oz.
  • Supplier No.:
    72236-50

Specifications

About this item

Finally, your automated coverslipper has the right coverslip to allow it to run smoothly. These “Superslip®” coverslips are designed to work in automatic coverslip machines and humid environments without sticking or jamming up in the middle of a run.

These new superslips combine specialized production procedures with our unique moisture-resistant packaging which brings you superior coverslips.

Customers Who Bought This Also Bought