Specifications
- Description:Diamond scriber, straight mount
- Cat. no.:100502-476
- Supplier No.:62108-ST
- Diameter:0.25 mm
Specifications
About this item
- Fine scribing under the microscope - for silicon wafers and glass coverslips
- Precision scribing and repairing - for thin film circuits and microcircuits, and more
This diamond scriber is the ultimate scribing instrument offering the most precision for the most delicate applications. It is available in straight or bent configurations.The integrated 6.35 mm anodized hexagonal (non-rolling) aluminum handle has a positive finger grip in the form of grooves for easy control and nonslippage. Each diamond scriber is packaged in a protective plastic tube.