Zerostat Anti-Static Gun, Electron Microscopy Sciences
Catalog #: 100496-120
Supplier: Electron Microscopy Science
Minority or Woman-Owned Business Enterprise
Specifications
- Description:Zero-Stat Anti-Static Gun
- Supplier No.:60610
Specifications
About this item
The Zerostat Anti-Static Gun operates on a piezoelectric effect to emit positive and negative charges to neutralize static on any surface. This anti-static gun emits no radioactivity and does not require electricity or batteries.
Measures: 4×6"
Customers Who Bought This Also Bought