To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- pH:—
- Description:Sorensen's Phosphate Buffer, 0.15 M
- Size:2 x 0.5 mm Slot
- Size:500 ml
- Volume:500 ml
- Supplier No.:11682-05
Specifications
About this item
Sorensen's Phosphate Buffer is also know as phosphate mixed sodium salts. The osmolarity of a 0.1 M at a pH 7.2 is 226 mosmols; with the addition of 0.18 M of sucrose to the solution, the osmolarity raises to 425 mosmols.
- pH can be specified when ordering
- Maximum shelf life after receipt is 3 months