ContactUsLinkComponent
Differential Blood Cell Counter
Blood Cell Counter, Bal Supply
Catalog #: 15173-273
Supplier:  Tip Industries
Differential Blood Cell Counter
Blood Cell Counter, Bal Supply
Catalog #: 15173-273
Supplier:  Tip Industries
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Counter with Two Counting Units
  • Cat. no.:
    15173-273
  • Supplier No.:
    42202C
Specifications
About this item
Counter with two counting units has single reset knob where as counter with five and eight counting units have reset knobs on each side.
Counting units have a range of 0 to 999. Removable tabs provide an area for identification of the classification being counted.
Customers Who Bought This Also Bought