Flash Column, Transparent, Alumina Acidic C-Series 40-60 ?M 60A 4G 20/Pack
Catalog #: 89216-250
Supplier: Agela Technologies
Flash Column, Transparent, Alumina Acidic C-Series 40-60 ?M 60A 4G 20/Pack
Catalog #: 89216-250
Supplier: Agela Technologies
Customers Who Bought This Also Bought
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
109,33 $ to 364,87 $
Puritan® UniTranz-RT® Media Transport Systems, Puritan Medical Products
Supplier: Puritan Medical Products
304,06 $ to 1 516,46 $