Flash Column Acid Aluminum 40-60 ?M 60A 120G 5/Pack
Catalog #: 89214-014
Supplier: Agela Technologies
Flash Column Acid Aluminum 40-60 ?M 60A 120G 5/Pack
Catalog #: 89214-014
Supplier: Agela Technologies
Customers Who Bought This Also Bought
VWR® Yttrium, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging
Supplier: VWR International
$848.41
Masterflex® Luer Stopcock Fittings, Polycarbonate, Avantor®
Supplier: VWR International
$64.62 to $66.68
Fluoride Standards, 1 ml = 1 mg F-, 1000 ppm F-, RICCA Chemical Company
Supplier: Ricca Chemical
$166.86
Starch 0.5% (w/v) in aqueous solution for iodometric titrations mercury-free
Supplier: Ricca Chemical
$96.29
Carbon Conductive Tape, Double Coated, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$261.44
Tryptic Soy Agar (TSA), Soybean Casein Digest Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
$39.00