Flash Column Acid Aluminum 40-60 ?M 60A 4G 20/Pack
Catalog #: 89214-004
Supplier: Agela Technologies
Flash Column Acid Aluminum 40-60 ?M 60A 4G 20/Pack
Catalog #: 89214-004
Supplier: Agela Technologies
Customers Who Bought This Also Bought
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$500.19 to $1,790.36
Ethanol denatured, anhydrous (denatured with methanol and ethyl acetate)
Supplier: INNOVA CHEMICALS CA
$468.00