Flash Column Base Aluminum 40-60 ?M 60A 1500G Each
Catalog #: 89214-002
Supplier: Agela Technologies
Flash Column Base Aluminum 40-60 ?M 60A 1500G Each
Catalog #: 89214-002
Supplier: Agela Technologies
Customers Who Bought This Also Bought
Sodium carbonate, anhydrous ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
129,84 $ to 1 081,96 $
Accessories for Advanced® Multi-Sample Osmometer, Model 2020, Advanced Instruments
Supplier: Advanced Instruments
1 223,38 $
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
260,09 $ to 2 400,95 $
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
881,80 $ to 2 807,56 $
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
458,88 $ to 1 237,78 $
di-Ammonium oxalate monohydrate, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
1 018,93 $ to 3 977,11 $