Flash Column Base Aluminum 40-60 ?M 60A 40G 10/Pack
Catalog #: 89213-992
Supplier: Agela Technologies
Flash Column Base Aluminum 40-60 ?M 60A 40G 10/Pack
Catalog #: 89213-992
Supplier: Agela Technologies
Customers Who Bought This Also Bought
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
$205.10 to $1,872.65
Water, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$277.09 to $740.95
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Alcohol ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
$131.08 to $405.43