Flash Column Base Aluminum 40-60 ?M 60A 12G 20/Pack
Catalog #: 89213-988
Supplier: Agela Technologies
Flash Column Base Aluminum 40-60 ?M 60A 12G 20/Pack
Catalog #: 89213-988
Supplier: Agela Technologies
Customers Who Bought This Also Bought
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
$109.33 to $364.87
Calcium carbonate 99.95-100.05% ACS chelometric standard
Supplier: Thermo Scientific Chemicals
$114.99
Choline chloride 98.0-100.5% (by anhydrous basis), BAKER ANALYZED®, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$149.28
Anti-CD41/CD61 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: PAC-1]
Supplier: Biolegend
$510.34