Flash Column Neutral Aluminum 40-60 ?M 60A 120G 5/Pack
Catalog #: 89213-978
Supplier: Agela Technologies
Flash Column Neutral Aluminum 40-60 ?M 60A 120G 5/Pack
Catalog #: 89213-978
Supplier: Agela Technologies
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
Masterflex® L/S® High-Performance Precision Tubing, Puri-Flex®, Avantor®
Supplier: VWR International
$158.00