Flash Column Neutral Aluminum 40-60 ?M 60A 4G 20/Pack
Catalog #: 89213-968
Supplier: Agela Technologies
Flash Column Neutral Aluminum 40-60 ?M 60A 4G 20/Pack
Catalog #: 89213-968
Supplier: Agela Technologies
Customers Who Bought This Also Bought
Sodium hydroxide 40% (w/w) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$146.85 to $576.51
Masterflex® Luer Stopcock Fittings, Polycarbonate, Avantor®
Supplier: VWR International
$64.62 to $66.68